Learn More
Invitrogen™ TIMP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA138778
Description
This antibody is predicted to react with rabbit and baboon based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is placenta or breast carcinoma.
Tissue inhibitor of metalloproteinase 3 (TIMP3) is a member of the tissue inhibitor of metalloproteinases gene family. It functions to inhibit the activity of matrix metalloproteinases (MMP) which degrade components of the extracellular matrix. TIMP3 is expressed upon mitogenic stimulation. It binds irreversibly to zinc-dependent MMPs and inactivates them by binding to their catalytic zinc cofactor. Other functions of TIMP3 include nervous system development, tissue regeneration, and visual perception. In humans, the gene encoding TIMP3 is located on chromosome 22.
Specifications
TIMP3 | |
Polyclonal | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
P35625 | |
TIMP3 | |
Synthetic peptide derived from the C-terminus of human TIMP-3. | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
TIMP3 | |
HSMRK222; K222; K222TA2; Metalloproteinase inhibitor 3; MIG-5 protein; protein MIG-5; SFD; Sun; TIMP 3; TIMP metallopeptidase inhibitor 3; TIMP metallopeptidase inhibitor 3 (Sorsby fundus dystrophy, pseudoinflammatory); TIMP metallopeptidase inhibitor 3 L homeolog; timp3; TIMP-3; TIMP-3 protein; timp3.L; timp3-A; tissue inhibitor metalloproteinase-3; tissue inhibitor of metalloproteinase 3; tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, pseudoinflammatory); tissue inhibitor of metalloproteinase-3; tissue inhibitor of metalloproteinase-3 precursor; tissue inhibitor of metalloproteinases 3; tissue inhibitor of metalloproteinases-3; XELAEV_18002484mg | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7078 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.