Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMP3 Recombinant Superclonal™ Antibody (20HCLC)

Catalog No. PI710404
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI710404 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI710404 Supplier Invitrogen™ Supplier No. 710404
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Superclonal Antibody

Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

Tissue inhibitor of metalloproteinase 3 (TIMP3) is a member of the tissue inhibitor of metalloproteinases gene family. It functions to inhibit the activity of matrix metalloproteinases (MMP) which degrade components of the extracellular matrix. TIMP3 is expressed upon mitogenic stimulation. It binds irreversibly to zinc-dependent MMPs and inactivates them by binding to their catalytic zinc cofactor. Other functions of TIMP3 include nervous system development, tissue regeneration, and visual perception. In humans, the gene encoding TIMP3 is located on chromosome 22.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMP3
Applications Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
Clone 20HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene TIMP3
Gene Accession No. P35625, P39876
Gene Alias HSMRK222; K222; K222TA2; Metalloproteinase inhibitor 3; MIG-5 protein; protein MIG-5; SFD; Sun; TIMP 3; TIMP metallopeptidase inhibitor 3; TIMP metallopeptidase inhibitor 3 (Sorsby fundus dystrophy, pseudoinflammatory); TIMP metallopeptidase inhibitor 3 L homeolog; timp3; TIMP-3; TIMP-3 protein; timp3.L; timp3-A; tissue inhibitor metalloproteinase-3; tissue inhibitor of metalloproteinase 3; tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, pseudoinflammatory); tissue inhibitor of metalloproteinase-3; tissue inhibitor of metalloproteinase-3 precursor; tissue inhibitor of metalloproteinases 3; tissue inhibitor of metalloproteinases-3; XELAEV_18002484mg
Gene Symbols TIMP3
Host Species Rabbit
Immunogen proprietary.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21859, 7078
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.