Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP4 Monoclonal Antibody (9:4-7), Rockland™
SDP

Catalog No. 501990596
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0596 100 μg
50-199-0597 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0596 Supplier Rockland Immunochemicals Supplier No. 200301E65
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. TIMP4 antibody is specific for human TIMP4 protein. This antibody reacts with human TIMP4 in tissue sections of formalin-fixed, paraffin-embedded In ELISAs and Native-PAGE, this antibody does not cross-react with recombinant human TIMP-1, -2 or -3. A BLAST analysis was used to suggest cross-reactivity with TIMP4 from human and mouse sources based on the immunizing sequence. Cross-reactivity with TIMP-4 from other sources has not been determined.

Tissue inhibitor of metalloproteinase 4 (TIMP4) is encoded by the gene TIMP4 located on chromosome 3. It belongs to the family of TIMP genes and is known to inhibit the activity of matrix metalloproteinases. Matrix metalloproteinases (MMP) are a group of proteinases that degrade the components of extracellular matrix. TIMP4 irreversibly binds to MMPs and inactivates them with the help of zinc as a cofactor. The netrin domain of TIMP4 is involved in platelet recruitment and aggregation. Other biological functions include response to cytokine and peptide hormone stimuli and involvement with the ovulation cycle and CNS development.

Specifications

Antigen TIMP4
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 9:4-7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene TIMP4
Gene Accession No. Q99727, Q9JHB3
Gene Alias Metalloproteinase inhibitor 4; TIMP metallopeptidase inhibitor 4; Timp4; TIMP-4; tissue inhibitor of metalloproteinase 4; Tissue inhibitor of metalloproteinases 4
Gene Symbols TIMP4
Host Species Mouse
Immunogen Anti-TIMP4 (MOUSE) Monoclonal Antibody was produced in mouse by repeated immunizations with full length human TIMP4 protein followed by hybridoma development.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110595, 7079
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.