Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMP4, Mouse anti-Human, Clone: 9:4-7, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16419
Specifications
TIMP4 | |
Monoclonal | |
Unconjugated | |
In 20 mM K3PO4, 150 mM NaCl, pH 7.2 (0.01% sodium azide). | |
- | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
9:4-7 | |
ELISA (1:20000-1:100000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:2000-1:10000) The optimal working dilution should be determined by the end user. | |
TIMP metallopeptidase inhibitor 4 | |
TIMP4 | |
Recombinant protein corresponding to full length human TIMP4 followed by hybridoma development. | |
RUO | |
7079 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction