Learn More
Invitrogen™ TIMP4 Polyclonal Antibody

Description
TIMP4 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | TIMP4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | TIMP4 |
| Gene Accession No. | Q99727, Q9JHB3 |
| Gene Alias | Metalloproteinase inhibitor 4; TIMP 4; TIMP metallopeptidase inhibitor 4; TIMP4; TIMP-4; tissue inhibitor of metalloproteinase 4; tissue inhibitor of metalloproteinases 4 |
| Gene Symbols | TIMP4 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.