Learn More
Invitrogen™ TIMP4 Recombinant Superclonal™ Antibody (13HCLC)
Rabbit Recombinant Superclonal Antibody
Supplier: Invitrogen™ 710340
Description
Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
Tissue inhibitor of metalloproteinase 4 (TIMP4) is encoded by the gene TIMP4 located on chromosome 3. It belongs to the family of TIMP genes and is known to inhibit the activity of matrix metalloproteinases. Matrix metalloproteinases (MMP) are a group of proteinases that degrade the components of extracellular matrix. TIMP4 irreversibly binds to MMPs and inactivates them with the help of zinc as a cofactor. The netrin domain of TIMP4 is involved in platelet recruitment and aggregation. Other biological functions include response to cytokine and peptide hormone stimuli and involvement with the ovulation cycle and CNS development.
Specifications
TIMP4 | |
Recombinant Superclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q99727 | |
TIMP4 | |
proprietary. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
13HCLC | |
Unconjugated | |
TIMP4 | |
Metalloproteinase inhibitor 4; TIMP 4; TIMP metallopeptidase inhibitor 4; TIMP4; TIMP-4; tissue inhibitor of metalloproteinase 4; tissue inhibitor of metalloproteinases 4 | |
Rabbit | |
Protein A | |
RUO | |
7079 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.