Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIN-Ag Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983100
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3100 Supplier Bioss Supplier No. BS13712R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mediates adhesion of proximal tubule epithelial cells via integrins alpha3-beta1 and alphaV-beta3. This is a non catalytic peptidase C1 family protein.

Specifications

Antigen TIN-Ag
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Tinag
Gene Accession No. Q9UJW2
Gene Alias AI452335; BOS_21298; LOC100153159; TINAG; TIN-Ag; Tubulointerstitial nephritis antigen; tubulointerstitial nephritis antigen-like
Gene Symbols Tinag
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TIN-Ag.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27283
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.