Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin bis(trifluoromethylsulfonyl)imide

Catalog No. AA4714206
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 4600096-08-6

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4600096-08-6
MDL Number MFCD28100488
Synonym Tin bis(trifluoromethanesulfonyl)imide; Tin bis(trifluoromethanesulfonimide)

Specifications

Color White
Linear Formula [(CF3SO2)2N]2Sn
Quantity 5 g
UN Number UN3261
Sensitivity Hygroscopic
Formula Weight 678.98
Physical Form Solid
Chemical Name or Material Tin bis(trifluoromethylsulfonyl)imide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.