Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tin foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.998% (metals basis)

Click to view available options
Quantity:
50 x 50mm
100 x 100mm
100 x 500mm
7440-31-5
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german
5352426
CHEBI:32990
tin
[Sn]
7440-31-5
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german
5352426
CHEBI:32990
tin
[Sn]
Quantity 50 x 50mm
Weight ≈1.8g/50x50mm
Odor Odorless
Physical Form Foil
Percent Purity 99.998%
Grade Puratronic™
Chemical Name or Material Tin foil, 0.1mm (0.004 in.) thick
Assay (metals basis)

EINECSNumber : 231-141-8

TSCA : Yes

Recommended Storage : Ambient temperatures