Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin ICP-MS Standard, 10,000 ppm Sn in 5% HNO3, Ricca Chemical
SDP

Catalog No. MSSN10KN100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSSN10KN-100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSSN10KN-100 Supplier Ricca Chemical Company Supplier No. MSSN10KN100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7440-31-5 , 7697-37-2 , 7664-39-3 , 7732-18-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 10,000ppm Sn
CAS Min % 84.0, 12.56, 1.04, 0.84
CAS Max % 87.0, 13.07, 1.09, 0.87
Equivalence 1 mL = 10 mg Sn
Physical Form Liquid
Color Colorless
pH < 2
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Single-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Tin ICP-MS Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.