Learn More
Tin(II) 2-ethylhexanoate, tech.
Description
Tin(II) 2-ethylhexanoate is used as a polymerization initiator in polylactic acid production. It acts as an intermediate as well as a catalyst for urethane foams, lubricants, addition agents and stabilizers for transformer oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 1.251 g/mL |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.494 |
| Quantity | 2 kg |
| Solubility Information | Miscible with water. |
| Formula Weight | 405.11 |
| Grade | Technical |
| Physical Form | Liquid |
| Chemical Name or Material | Tin(II) 2-ethylhexanoate |
Safety and Handling
| Hazard Category | H315-H319-H335-H361d |
| Hazard Statement | GHS H Statement H361-H315-H319-H335 Suspected of damaging fertility or the unborn child. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c |
| EINECSNumber | 206-108-6 |
| RTECSNumber | MO7870000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.