Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(II) acetate, 95%, Alfa Aesar™

Manufacturer:  Alfa Aesar 2236114

Quantity 25g

 View more versions of this product

Catalog No. AA2236114


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

638-39-1
95%
236.8
Stannous acetate
Powder
Sn(OOCCH3)2
MFCD00054408
Tin(II) acetate

Specifications

638-39-1
95%
White powder
MFCD00054408
2.31
25g
Powder
Sn(OOCCH3)2
236.8
Stannous acetate
White
Tin(II) acetate

Safety and Handling

SDS & Safety Documents

H331-H302-H312: Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 211-335-9

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit