Tin(II) acetate, 95%, Alfa Aesar™

Manufacturer: Alfa Aesar 2236106

Quantity 5g

 View more versions of this product

Catalog No. AA2236106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 638-39-1
    Physical Form Powder
    Assay Percent Range 95%
    Molecular Formula Sn(OOCCH3)2
    Formula Weight 236.8
    MDL Number MFCD00054408
    Synonym Stannous acetate
    Chemical Name or Material Tin(II) acetate

    Specifications

    Physical Form Powder
    Formula Weight 236.8
    Quantity 5g
    Color White
    DOT Information DOT Class: 6.1, Packaging Group: III
    EINECS Number 211-335-9
    Appearance White powder
    CAS 638-39-1
    Assay Percent Range 95%
    Molecular Formula Sn(OOCCH3)2
    MDL Number MFCD00054408
    Synonym Stannous acetate
    Health Hazard 1 H331-H302-H312: Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin.
    Chemical Name or Material Tin(II) acetate

    H331-H302-H312: Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit