Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(II) chloride dihydrate, 98%

Catalog No. aaa1461022
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 10025-69-1 | Cl2Sn·2H2O

Tin(II) chloride is used as a sensitizing agent in preparing glass and plastic for metalizing. It serves as a potent reducing agent. Other uses of stannous chloride are tin electroplating baths, corrosion inhibitors, polymers, thermoplastic elastomers, soldering flux, antioxidant, tanning agent and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tin(II) chloride is used as a sensitizing agent in preparing glass and plastic for metalizing. It serves as a potent reducing agent. Other uses of stannous chloride are tin electroplating baths, corrosion inhibitors, polymers, thermoplastic elastomers, soldering flux, antioxidant, tanning agent and pharmaceuticals.

Solubility
Soluble in dilute and concentrated hydrochloric acid, alcohol, ethyl acetate, glacial acetic acid, and sodium hydroxide solution. Decomposes in excess water.

Notes
Hygroscopic, and absorbs moisture or water from air. Incompatible with metals, strong oxidizing agents, strong bases and bromine trifluoride.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2Sn·2H2O
MDL Number MFCD00149863
Synonym Tin (2+) dihydrate dichloride
IUPAC Name Tin (2+) dihydrate dichloride

Specifications

Melting Point 42°C to 46°C
Quantity 100 g
Linear Formula SnCl2·2H2O
UN Number UN3260
Merck Index 14,8783
Solubility Information Soluble in dilute and concentrated hydrochloric acid,alcohol,ethyl acetate,glacial acetic acid,and sodium hydroxide solution. Decomposes in excess water.
Formula Weight 225.65 (189.62 Anhydrous)
Percent Purity 98%
Sensitivity Air Sensitive
Density 2.71
Chemical Name or Material Tin(II) chloride dihydrate
Show More Show Less
Hazard Category H290-H314-H317-H332-H335-H373
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P234-P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 600-045-1
RTECSNumber XP8850000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.