Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(II) chloride dihydrate, For AAS, ≥96.0% (RT), MilliporeSigma™ Supelco™

Catalog No. 111018458 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8458 250 g
11-101-9665 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8458 Supplier MilliporeSigma Supelco Supplier No. 96527250G
Add to Cart
Add to Cart

Tin(II) chloride dehydrate converts conjugated dioxolones to aldehyde. It helps as a catalyst in multi-component condensation reaction from aromatic aldehydes, 2-aminopyridines and isonitriles which results in reasonable yield and reducing the reaction time.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2H4O2Sn
Molecular Weight (g/mol) 225.64
MDL Number MFCD00149863
InChI Key FWPIDFUJEMBDLS-UHFFFAOYSA-L
Synonym Stannous chloride dihydrate
IUPAC Name λ²-tin(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Sn++]

Specifications

Linear Formula SnCl2 · 2H2O
Melting Point 37°C to 38°C (decomposition)
Boiling Point 652°C
Quantity 250 g
Formula Weight 225.65
Percent Purity ≥96.0% (RT)
Grade AAS
RTECSNumber XP8850000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.