Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(Ii) Chloride Dihydrate, puriss. p.a. ACS Reagent, Solstice

Catalog No. 6001695 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-016-95 100 g
60-016-97 500 g
60-016-96 1 kg
60-045-971 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-016-95 Supplier Solstice Supplier No. 31669100G

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

Puriss. p.a., ACS Reagent, Reagent ISO, Reagent European Pharmacopoeia,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-69-1
MDL Number MFCD00149863

Specifications

Quantity 100 g
Linear Formula SnCl2 · 2H2O
UN Number 3260
Formula Weight 225.65g/mol
Percent Purity ≥98%
Grade ACS/puriss. p.a.
Chemical Name or Material Tin(Ii) Chloride Dihydrate, Reag. Ph. Eur. reag. ISO
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H314-H317-H332-H335-H341-H361-H373-H410
EINECSNumber 231-868-0
RTECSNumber XP8850000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.