Learn More
Tin(II) chloride dihydrate, Reagent Grade, Thermo Scientific Chemicals
Description
Stannous chloride is used as a sensitizing agent in preparing glass and plastic for metalizing. It serves as a potent reducing agent. Other uses of stannous chloride are tin electroplating baths, corrosion inhibitors, polymers, thermoplastic elastomers, soldering flux, antioxidant, tanning agent and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 43°C to 46°C |
Physical Form | Crystalline |
Quantity | 500 g |
Solubility Information | Soluble in dilute and concentrated HCl,alcohol,ethyl acetate,glacial acetic acid,and NaOH solution. Decomposes in excess water. |
Formula Weight | 225.65 (189.62 Anhydrous) |
Grade | Reagent |
Sensitivity | Air Sensitive |
Density | 2.71 |
Chemical Name or Material | Tin (II) Chloride dihydrate, Reagent Grade |
Safety and Handling
Hazard Category | H290-H314-H317-H332-H335-H373 |
Precautionary Statement | P234-P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P390-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: III |
EINECSNumber | 600-045-1 |
RTECSNumber | XP8850000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.