Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tin(II) ethoxide

Manufacturer:  Alfa Aesar™ 2089603

Quantity 1g

 View more versions of this product

Catalog No. AA2089603


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

14791-99-2
208.81
Tin(II) ethoxide
Sn(OC2H5)2
MFCD00015653

Specifications

≈200°C (dec.)
1g
Solid/Semi-Solid
208.81
Reacts with water
14791-99-2
Air & Moisture Sensitive
Sn(OC2H5)2
MFCD00015653
Tin(II) ethoxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit