Learn More
Tin(II) iodide, 99+%
Description
Tin(II) iodide is used to prepare organic-inorganic perovskite thin films which exhibit uniform surfaces and strong photoluminescence. It finds application as an analytical reagent in the determination of rhodium and iridium by spectrophotometry. It is also involved in the synthesis of CH3(CH2)11NH3]SnI3, which is a hybrid semiconductor with MoO3 type tin(II) iodide layers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 720°C (decomposition) |
Melting Point | 320°C |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | >99% |
Solubility Information | Soluble in benzene,carbon disulfide,chloroform,and solutions of alkali chlorides and iodides. Decomposes in water |
Formula Weight | 372.5 |
Sensitivity | Air, light, and moisture sensitive |
Density | 5.28 |
Chemical Name or Material | Tin(II) iodide |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | DOT Class: 8; Packaging Group: III |
EINECSNumber | 233-667-3 |
RTECSNumber | XQ3650000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.