Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Tin(II) Sulfate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 g
500 g
Description
General applications:Tin(II) sulfate can be used in the electrosynthesis of Cu2ZnSnS4 (CZTS) thin films for solar cell application.It can be used to synthesize ternary Cu-Sn-S chalcogenides such as Cu2SnS3, Cu5Sn2S7 and Cu3SnS4 by successive ionic layer adsorption and reaction (SILAR) method.SnSO4 can also be used in the preparation of tin (IV) oxide (SnO2) nanoparticles which are widely used in battery electrodes, fabrication of supercapacitors and opto-electronic devices.
Specifications
Specifications
| CAS | 7488-55-3 |
| Molecular Formula | O4SSn |
| Linear Formula | SnSO4 |
| MDL Number | MFCD00011246 |
| Quantity | 100 g |
| Synonym | Stannous sulfate |
| Molecular Weight (g/mol) | 214.77 g/mol |
| Biological Activity | Cardiovascular System, Respiratory System |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction