Alfa Aesar™ Tin(II) tetrafluoroborate, 50% w/w aq. soln.

Manufacturer: Alfa Aesar™ 89669D5

Quantity 3 x 1kg

 View more versions of this product

Catalog No. AA89669D5

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13814-97-6
    Molecular Formula B2F8Sn
    Linear Formula Sn(BF4)2
    Formula Weight 292.3
    MDL Number MFCD00054047
    UN Number UN3264
    Chemical Name or Material Tin (II) tetrafluoroborate

    Specifications

    Formula Weight 292.3
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Density 1.670g/mL
    Molecular Formula B2F8Sn
    Linear Formula Sn(BF4)2
    MDL Number MFCD00054047
    UN Number UN3264
    Color Colorless
    Concentration 50% w/w aq. soln.
    TSCA TSCA
    Physical Form Liquid
    CAS 13814-97-6
    Quantity 3 x 1kg
    Health Hazard 2 GHS H Statement
    H314
    Causes severe skin burns and eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
    Chemical Name or Material Tin (II) tetrafluoroborate

    GHS H Statement
    H314
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit