Learn More
Tin(II) tetrafluoroborate, 50% w/w aq. soln.
Description
Tin(II) tetrafluoroborate is used in electroplating. It is also used for plating bath tinned copper, tin and tin alloy. Further, it acts as a catalyst for various organic reactions. It is also employed in metal plating. In addition to this, it is used as a petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 1 kg |
| Linear Formula | Sn(BF4)2 |
| UN Number | UN3264 |
| Solubility Information | Miscible with water. |
| Formula Weight | 292.3 |
| Concentration or Composition (by Analyte or Components) | 50% w/w aq. soln. |
| Density | 1.670 g/mL |
| Chemical Name or Material | Tin (II) tetrafluoroborate |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314 Causes severe skin burns and eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S. |
| EINECSNumber | 237-487-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.