Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(IV) acetate

Catalog No. p-4782191
Click to view available options
Quantity:
5 g
25 g

CAS: 2800-96-6 | C8H12O8Sn | 354.886 g/mol

Tin(IV) acetate is used to prepare tin-based oxides (Sn-Zn-P-O), which is a negative electrodes for rechargeable lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2800-96-6
Molecular Formula C8H12O8Sn
Molecular Weight (g/mol) 354.886
MDL Number MFCD00014976
InChI Key YJGJRYWNNHUESM-UHFFFAOYSA-J
Synonym tin iv acetate, tin acetate, tetraacetoxystannane, tin-iv-acetate, acmc-20ajhv, tin 4+ ion tetraacetate, tetraacetoxytin, tin 4+ tetraacetate
PubChem CID 9863446
IUPAC Name tin(4+);tetraacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sn+4]

Specifications

Melting Point 232°C to 233°C
Physical Form Crystalline
Quantity 5 g
Solubility Information Soluble in acetone and benzene.
Formula Weight 354.89
Sensitivity Hygroscopic
Chemical Name or Material Tin(IV) Acetate
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.