Alfa Aesar™ Tin(IV) bromide, 99%

Manufacturer: Alfa Aesar™ 7112018

Quantity 50g

 View more versions of this product

Catalog No. AA7112018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-67-5
    Molecular Formula SnBr4
    Formula Weight 438.33
    MDL Number MFCD00011240
    UN Number UN3260
    Merck Index 14,8773
    Chemical Name or Material Tin (IV) bromide

    Specifications

    Physical Form Lumps
    Formula Weight 438.33
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Acidic
    Sensitivity Air & Moisture Sensitive
    Merck Index 14,8773
    Quantity 50g
    Health Hazard 2 GHS H Statement
    H314-H318-H335
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause respiratory irritation.
    CAS 7789-67-5
    Boiling Point 202°C (734mmHg)
    Melting Point 31°C
    Health Hazard 1 Danger
    Density 3.34g/mL
    RTECS Number XP8300000
    Molecular Formula SnBr4
    MDL Number MFCD00011240
    UN Number UN3260
    TSCA TSCA
    Color Colorless to white
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    Chemical Name or Material Tin (IV) bromide
    Assay 99%

    GHS H Statement
    H314-H318-H335
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause respiratory irritation.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit