Tin(IV) chloride, 99.99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 7120322

Quantity 100g

 View more versions of this product

Catalog No. AA7120322

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7646-78-8
    Physical Form Liquid
    Assay Percent Range 99.99% (metals basis)
    Molecular Formula Cl4Sn
    Formula Weight 260.5
    MDL Number MFCD00011242
    Synonym Stannic chloride
    Chemical Name or Material Tin(IV) chloride

    Specifications

    Formula Weight 260.5
    Quantity 100g
    EINECS Number 231-588-9
    Physical Form Liquid
    Assay Percent Range 99.99% (metals basis)
    Molecular Formula Cl4Sn
    MDL Number MFCD00011242
    CAS 7646-78-8
    Synonym Stannic chloride
    Chemical Name or Material Tin(IV) chloride

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit