Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(IV) isopropoxide isopropanol adduct, 98% (metals basis)

Catalog No. aa4126903
Click to view available options
Quantity:
1 g
5 g

CAS: 132951-93-0

Tin(IV) isopropoxide isopropanol adduct is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, electrochemical and conducting properties and thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132951-93-0
MDL Number MFCD00145408

Specifications

Melting Point ∼130°C (decomposition)
Boiling Point 131°C (1.6 mmHg)
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Soluble in hydrocarbons and warm isopropanol. Hydrolyzes with water. Reacts with alcohols,ketones and esters.
Formula Weight 414.15
Percent Purity 98%
Physical Form Wet Solid
Assay (metals basis)
Chemical Name or Material Tin(IV) isopropoxide isopropanol adduct
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.