Tin selenide, 99.999% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1878114

Quantity 25g

 View more versions of this product

Catalog No. AA1878114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1315-06-6
    Physical Form 3 to 12mm Pieces
    Assay Percent Range 99.999% (metals basis)
    Molecular Formula SnSe
    Formula Weight 197.65
    MDL Number MFCD00054049
    Chemical Name or Material Tin selenide

    Specifications

    Physical Form 3 to 12mm Pieces
    Formula Weight 197.65
    Quantity 25g
    EINECS Number 215-257-6
    Molecular Formula SnSe
    Assay Percent Range 99.999% (metals basis)
    MDL Number MFCD00054049
    CAS 1315-06-6
    Chemical Name or Material Tin selenide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit