Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™
SDP

Catalog No. orgsn82z Shop All Spex CertiPrep Products
Click to view available options
Quantity:
50 g

Catalog No. ORG-SN8-2Z

Supplier: SPEX CertiPrep ORGSN82Z

Only null left
Add to Cart
Add to Cart

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sn1000μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7440-31-5, 8042-47-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Packaging Triple-leached LDPE bottle
Certificate of Analysis With Certificate of Analysis.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Min % 0.5, 99.5
CAS Max % 0.5, 99.5
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Base Oil 20
Industry Type Petrochemical Testing
Quantity 50 g
Grade Oil/Plasma
Concentration or Composition (by Analyte or Components) Matrix Concentration Cat. No.
Base Oil 1000μg/g ORG-SN8-2Z
Base Oil 5000μg/g ORG-SN8-4Z
Chemical Name or Material Tin Oil Standard
Show More Show Less
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.