Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin Standard, 1mL = 1mg Sn, 1000ppm Sn, Ricca Chemical
SDP

Catalog No. 85004 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Poly natural

Catalog No. 85004

Supplier: Ricca Chemical Company 85004

Chemical Identifiers

CAS 7440-31-5, 7647-01-0, 7732-18-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Packaging Poly natural
Concentration or Composition Notes 1,000ppm Sn
Equivalence 1 mL = 1 mg Sn
Physical Form Liquid
Color Colorless
Quantity 120 mL
Chemical Name or Material Tin Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.