Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIN2 Antibody (5G11), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315221
Description
TIN2 Monoclonal antibody specifically detects TIN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
TIN2 | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4), 150mM NaCl, and 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
26277 | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA | |
5G11 | |
Western Blot 1:500-1:5000, ELISA | |
(TRF1)-interacting nuclear factor 2 variant 1, TERF1 (TRF1)-interacting nuclear factor 2, TERF1-interacting nuclear factor 2, TIN2TRF1-interacting nuclear protein 2 | |
A synthesized peptide derived from human TIN2. UniProt ID (Q9BSI4). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction