Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIN2 Antibody (SR1681), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | TIN2 |
---|---|
Clone | SR1681 |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
TIN2 Monoclonal antibody specifically detects TIN2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
TIN2 | |
Western Blot 1:500-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
26277 | |
IgG | |
Affinity purified |
SR1681 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
(TRF1)-interacting nuclear factor 2 variant 1, TERF1 (TRF1)-interacting nuclear factor 2, TERF1-interacting nuclear factor 2, TIN2TRF1-interacting nuclear protein 2 | |
A synthesized peptide derived from human TIN2 (Uniprot #: Q9BSI4) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title