Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111477
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111477 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111477 Supplier Invitrogen™ Supplier No. PA5111477
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRVQSQWAST RGGHKERPTV MLFPFRNLGS PTQVISKPES KEEHAIYTAD LAMGTRAAST GKSKSPCQTL GGRALKENPV DL Highest antigen sequence indentity to the following orthologs: Mouse - 48%, Rat - 54%.

TRF1 is a negative regulator of telomere length maintenance. Recently two proteins, tankyrase and Tin2, which bind to mammalian TRF1, have been identified. Tankyrase is a protein with homology to ankyrins as well as to the catalytic domain of poly (adenosine diphosphate-ribose) polymerase (PARP). Tankyrase localizes to telomeres by binding to the telomeric repeat binding factor 1 (TRF1) through its ankyrin repeats. Tankyrase exhibits PARP activity functioning as acceptors for adenosine diphosphate (ADP)-ribosylation. Since ADP-ribosylation of TRF1 diminishes its ability to bind to telomeric DNA, this suggests that telomere function in human cells is regulated by poly (ADP)-ribosylation. Both the cell cycle and TRF1 may regulate the subcellular localization of tankyrase. The cDNA coding for TIN2 protein was isolated as interacting partner of TRF1 from a yeast two-hybrid cDNA library screening. Tin2 localizes to telomeres and is essential for proper regulation of telomere length. TIN2, like TRF1, is widely and constitutively expressed, suggesting that these proteins act together to counterbalance telomere elongation by telomerase. However, endogenous TIN2, like endogenousTRF1, is not highly expressed, and therefore may be difficult to visualize by immunostaining in certain cell systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIN2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tinf2
Gene Accession No. Q9BSI4
Gene Alias AW552114; D14Wsu146e; DKCA3; TERF1 (TRF1)-interacting nuclear factor 2; TERF1 interacting nuclear factor 2; TERF1-interacting nuclear factor 2; tin; TIN2; Tin-2; TINF2; TRF1-interacting nuclear factor 2; TRF1-interacting nuclear protein 2
Gene Symbols Tinf2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TIN2. Recombinant protein control fragment (Product #RP-106912).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26277
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.