Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TINAGL1 Polyclonal Antibody
TINAGL1 Polyclonal Antibody
Supplier: Thermo Scientific PA589001
Description
TINAGL1 Polyclonal Antibody for Western Blot

Specifications
TINAGL1 | |
Polyclonal | |
Unconjugated | |
TINAGL1 | |
1110021J17Rik; Adrenocortical zonation factor 1; androgen-regulated gene 1; androgen-regulated gene 1 protein; Arg1; AZ1; AZ-1; Gis5; glucocorticoid-inducible protein 5; LCN7; LIECG3; lipocalin 7; OLRG2; OLRG-2; Oxidized LDL-responsive gene 2 protein; oxidized-LDL responsive gene 2; P3ECSL; PP6614; PSEC0088; TARP; TIN Ag-related protein; Tinagl; TINAGL1; TINAG-like 1; TINAGRP; TIN-Ag-RP; tubulointerstitial nephritis antigen like 1; tubulointerstitial nephritis antigen-like; tubulointerstitial nephritis antigen-like 1; tubulointerstitial nephritis antigen-related protein; UNQ204/PRO230 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
64129, 94174, 94242 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.79 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q99JR5, Q9EQT5, Q9GZM7 | |
TINAGL1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 258-467 of human TINAGL1 (NP_0714471). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction