Learn More
Invitrogen™ TINAGL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114111
Description
Immunogen sequence: NLIHEPLDQG NCAGSWAFST AAVASDRVSI HSLGHMTPVL SPQNLLSCDT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

Specifications
TINAGL1 | |
Polyclonal | |
Unconjugated | |
TINAGL1 | |
1110021J17Rik; Adrenocortical zonation factor 1; androgen-regulated gene 1; androgen-regulated gene 1 protein; Arg1; AZ1; AZ-1; Gis5; glucocorticoid-inducible protein 5; LCN7; LIECG3; lipocalin 7; OLRG2; OLRG-2; Oxidized LDL-responsive gene 2 protein; oxidized-LDL responsive gene 2; P3ECSL; PP6614; PSEC0088; TARP; TIN Ag-related protein; Tinagl; TINAGL1; TINAG-like 1; TINAGRP; TIN-Ag-RP; tubulointerstitial nephritis antigen like 1; tubulointerstitial nephritis antigen-like; tubulointerstitial nephritis antigen-like 1; tubulointerstitial nephritis antigen-related protein; UNQ204/PRO230 | |
Rabbit | |
Affinity chromatography | |
RUO | |
64129 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9GZM7 | |
TINAGL1 | |
Synthetic peptide directed towards the middle region of human TINAGL1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.