Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tinagl1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980917
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0917 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0917 Supplier Bioss Supplier No. BS10505R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen tinagl1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TINAGL1
Gene Accession No. Q9GZM7
Gene Alias 1110021J17Rik; Adrenocortical zonation factor 1; androgen-regulated gene 1; Androgen-regulated gene 1 protein; Arg1; AZ1; AZ-1; Gis5; Glucocorticoid-inducible protein 5; Lcn7; LIECG3; lipocalin 7; OLRG2; OLRG-2; Oxidized LDL-responsive gene 2 protein; oxidized-LDL responsive gene 2; P3ECSL; PP6614; PSEC0088; TARP; TIN Ag-related protein; Tinagl; Tinagl1; TINAG-like 1; TINAGRP; TIN-Ag-RP; tubulointerstitial nephritis antigen like 1; Tubulointerstitial nephritis antigen-like; tubulointerstitial nephritis antigen-like 1; Tubulointerstitial nephritis antigen-related protein; UNQ204/PRO230
Gene Symbols TINAGL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human tinagl1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64129
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.