Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tingley™ Rubber Work High-top Galoshes

Catalog No. p-4910018 Shop All Tingley Rubber Co. Products
Click to view available options
Size:
2X-Large, 12.5 to 14
3X-Large, 14.5 to 15
Large, 9.5 to 11
Medium, 8 to 9.5
Small, 6 to 7.5
X-Large, 11 to 12.5
This item is not returnable. View return policy
This item is not returnable. View return policy

Won't crack or stiffen, even in cold weather

  • Made from an exclusive stretch-rubber compound
  • Protect against alcohol, and many organic acids, salts and alkalis
  • No hardware to tear out or conduct electricity
  • No lining for easy decontamination
  • Deep, slip-resistant treads
  • Black

Specifications

Closure Type Button
Color Black
For Use With (Application) Food processing, transportation, utilities, construction, general industry, municipalities, and agriculture
Material Rubber
Size Medium, 8 to 9.5
Sole Features Deep-cleated outsole
Style High top galoshes
Resistant To Chemicals, acids, alcohols
Waterproof Yes
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.