Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tingley™ Workbrutes™ PVC Knee-High Overshoes

These PVC overshoes have thicker and deeper cleats for a better grip and longer wear

$53.00

Specifications

Color Black
Height (English) 14 in.
Material PVC
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-999-342A
View Documents
Tingley™
35141.LG
Large, 11 to 13
Pair for $53.00
Only null left
Add to Cart
 
19-999-342C
View Documents
Tingley™
35141.SM
Small, 6.5 to 8
Pair for $53.00
Only null left
Add to Cart
 
Description

Description

  • Injection-molded, seamless construction for 100% waterproof construction
  • Injection molding puts less material in the upper, eliminating unnecessary weight, for reduced leg fatigue
  • More material is added to the sole at the heel, toe and cleats for longer life
  • Thicker tread has deeper cleats with sharper edges for more traction
  • Key interior entry points are specially textured to enable overshoes to easily slip on and off over rubber sole shoes
  • No-pinch molded-in button and two button holes provide a snug fit and protection
  • Resistant to fats, certain acids, hydrocarbons and caustics

Food processing, construction, general industry, transportation and hazardous waste cleanup

Specifications

Specifications

Black
14 in.
PVC
Videos
SDS
Documents

Documents

Product Certifications

Not recommended for use in ketones, aldehydes and many solvents.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.