Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azithromycin USP Dihydrate, Medisca™
SDP

Catalog No. 18048142 Shop All Medisca Products
Click to view available options
Quantity:
25 g
100 g
500g
1 kg

Chemical Identifiers

CAS 117772-70-0

Specifications

Physical Form Powder
Color White
Quantity 500g
Percent Purity 945 to 1030 μg/mg
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.