Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIP120A Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149918AF488
Description
TIP120A Polyclonal antibody specifically detects TIP120A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TIP120A | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
cullin-associated and neddylation-dissociated 1, Cullin-associated and neddylation-dissociated protein 1, cullin-associated NEDD8-dissociated protein 1, DKFZp434M1414, FLJ90441, KIAA0829FLJ10114, p120 CAND1, TBP interacting protein, TBP-interacting protein 120A, TBP-interacting protein of 120 kDa A, TIP120AFLJ38691, TIP120FLJ10929 | |
The immunogen for this product maps to a region between residue 1180 and 1230 of human Cullin-Associated and Neddylation-Dissociated 1 using the numbering given in entry NP_060918.2 (GeneID 55832). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55832 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction