Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIP120B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TIP120B |
---|---|
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TIP120B Polyclonal specifically detects TIP120B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TIP120B | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
O75155 | |
23066 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MPVLVSGIIFSLADRSSSSTIRMDALAFLQGLLGTEPAEAFHPHLPILLPPVMACVADSFYKIAAEALVVLQELVRALWPLHRPRMLDPEPYVGEMSAVTLARLRATDLDQEVKERAISCMGH | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
cullin-associated and neddylation-dissociated 2 (putative), Cullin-associated and neddylation-dissociated protein 2, cullin-associated NEDD8-dissociated protein 2, epididymis tissue protein Li 169, KIAA0667TBP-interacting protein of 120 kDa B, p120 CAND2, TBP interacting protein, TBP-interacting protein 120B, TIP120BTp120b | |
CAND2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title