Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIP120B Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501566756
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6756 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6756 Supplier Bethyl Laboratories, Inc Supplier No. A304046AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

CAND2 may play a role in the assembly of ubiquitin ligase complexes and modulate the ubiquitination of target proteins. And it may be also a transcription regulator.

Specifications

Antigen TIP120B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CAND2
Gene Accession No. O75155, Q6ZQ73
Gene Alias cullin-associated and neddylation-dissociated 2 (putative), cullin-associated and neddylation-dissociated protein 2, epididymis tissue protein Li 169, KIAA0667, p120 CAND2, TBP interacting protein, TBP-interacting protein 120B, TBP-interacting protein of 120 kDa B, TIP120B, Tp120b
Gene Symbols CAND2
Host Species Rabbit
Immunogen Between 1186 and 1236
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23066, 67088
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.