Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIP30 Antibody (5D6), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259482UV
This item is not returnable.
View return policy
Description
TIP30 Monoclonal antibody specifically detects TIP30 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
TIP30 | |
Monoclonal | |
DyLight 350 | |
30 kDa HIV-1 TAT-interacting protein, CC3oxidoreductase HTATIP2, EC 1.1.1, EC 1.1.1.-, FLJ26963, HIV-1 Tat interactive protein 2, 30 kDa, HIV-1 Tat interactive protein 2, 30kDa, HIV-1 TAT-interactive protein 2, SDR44U1, Tat-interacting protein (30kD), TIP30short chain dehydrogenase/reductase family 44U, member 1 | |
Recombinant human TIP30 (1-242aa) purified from E. coli | |
0.1 mL | |
Apoptosis | |
10553 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, ELISA, Immunofluorescence | |
5D6 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction