Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIP30 Mouse anti-Human, Clone: 5D6, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP25948250UL
Description
TIP30 Monoclonal specifically detects TIP30 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| TIP30 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1079, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| 30 kDa HIV-1 TAT-interacting protein, CC3oxidoreductase HTATIP2, EC 1.1.1, EC 1.1.1.-, FLJ26963, HIV-1 Tat interactive protein 2, 30 kDa, HIV-1 Tat interactive protein 2, 30kDa, HIV-1 TAT-interactive protein 2, SDR44U1, Tat-interacting protein (30kD), TIP30short chain dehydrogenase/reductase family 44U, member 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 5D6 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| HTATIP2 | |
| Recombinant human HTATIP2 (1-242aa) purified from E. coli | |
| 50 μL | |
| Apoptosis | |
| 10553 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction