Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIP30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582247 Supplier Invitrogen™ Supplier No. PA582247
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALSKLREDFR MQNKSVFILG ASGETGRVLL KEILEQGLFS KVTLIGRRKL TFDEEAYKNV NQEVVDFEKL DDYASAFQGH DVGFC Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 92%.

CC3 (HTATIP2) is a member of the short-chain dehydrogenases/reductases (SDR) family. It is a novel serine/threonine kinase that phosphorylates the C-terminal domain (CTD) of the largest RNA polymerase II subunit and induces the expression of apoptosis related genes Bad and Siva, as well as metastasis suppressor NM23-H2. It also interacts with an estrogen receptor alpha-interacting coactivator CIA and regulates ERalpha-mediated c-myc transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIP30
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HTATIP2
Gene Accession No. Q9BUP3
Gene Alias 30 kDa HIV-1 TAT-interacting protein; AW111545; CC3; FLJ26963; HIV-1 Tat interactive protein 2; HIV-1 tat interactive protein 2, 30 kDa homolog; HIV-1 Tat interactive protein 2, 30kDa; HIV-1 tat interactive protein 2, homolog; HIV-1 tat interactive protein 2, homolog (human); HIV-1 TAT-interactive protein 2; Htatip2; oxidoreductase HTATIP2; SDR44U1; short chain dehydrogenase/reductase family 44U, member 1; Tat-interacting protein (30kD); tat-interacting protein 30 kDa; Tip30
Gene Symbols HTATIP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HTATIP2. Recombinant protein control fragment (Product #RP-89324).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10553
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.