Learn More
Tip41 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95
Specifications
| Antigen | Tip41 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-155-8155
|
Bethyl Laboratories, Inc
A300662AT |
20 μL |
Each of 1 for $167.95
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
TIPRL is an inhibitory regulator of protein phosphatase-2A .Specifications
| Tip41 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| TIPRL; TIP41, TOR signaling pathway regulator-like (S. cerevisiae); MGC3794; TIP; TIP41; dJ69E11.3; CG9578-like; OTTHUMP00000032581; OTTHUMP00000032582; TIP41, TOR signalling pathway regulator-like; putative MAPK activating protein; yeast YPR037W and worm C02C2.6 predicted proteins-like | |
| Between 200 and 250 | |
| Primary | |
| 4° C | |
| TIPRL |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| O75663, Q8BH58 | |
| 226591, 261726 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.