Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tip41 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501558156
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-8156 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-8156 Supplier Bethyl Laboratories, Inc Supplier No. A300663A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.

TIPRL is an inhibitory regulator of protein phosphatase-2A .

Specifications

Antigen Tip41
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TIPRL
Gene Accession No. O75663, Q8BH58
Gene Alias TIPRL; TIP41, TOR signaling pathway regulator-like (S. cerevisiae); MGC3794; TIP; TIP41; dJ69E11.3; CG9578-like; OTTHUMP00000032581; OTTHUMP00000032582; TIP41, TOR signalling pathway regulator-like; putative MAPK activating protein; yeast YPR037W and worm C02C2.6 predicted proteins-like
Gene Symbols TIPRL
Host Species Rabbit
Immunogen Between 225 and C-term
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226591, 261726
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.