Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIP47 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597944
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597944 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597944 Supplier Invitrogen™ Supplier No. PA597944
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TIP47 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

TIP47 has been described as cargo protein involved in the trafficking of the mannose-6-phosphate receptor between endosomes and the Golgi complex. Mannose 6-phophate receptors (MPRs) deliver lysosomal hydrolase from the Golgi to endosomes and then return to the Golgi complex. The protein encoded by this gene interacts with the cytoplasmic domains of both cation-independent and cation-dependent MPRs and is required for endosome to Golgi transport. This protein also binds directly to the GTPase RAB9, a member of the RAS oncogene family. The interaction with RAB9 has been shown to increase the affinity of this protein for its cargo. TIP47 has been localized in milk fat globule membranes of human and bovine origin. It has been described as a placental protein, as well. Increased amounts of TIP47 are secreted into circulation of cervix carcinoma patients. Therefore, this protein is probably significant as an oncodevelopmental marker.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIP47
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PLIN3
Gene Accession No. O60664
Gene Alias 1300012C15Rik; 47 kDa mannose 6-phosphate receptor-binding protein; 47 kDa MPR-binding protein; 47 kDa tail interaction protein; cargo selection protein TIP47; M6prbp1; mannose 6 phosphate receptor binding protein 1; mannose-6-phosphate receptor binding protein 1; mannose-6-phosphate receptor-binding protein 1; MGC11117; MGC2012; perilipin 3; perilipin-3; Placental protein 17; Plin3; PP17; tail-interacting protein, 47 kD; testicular tissue protein Li 114; TIP47; Unknown (protein for MGC:128897)
Gene Symbols PLIN3
Host Species Rabbit
Immunogen Recombinant Human Perilipin-3 protein (192-324AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10226
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.