Learn More
Invitrogen™ TIP47 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA146160
Description
In Western Blot analysis, a band is seen at ~47 kDa, representing isoform B. In some samples, a ~28 kDa band may be observed which represents a splice isoform. Suggested positive control: antigen standard for M6PRBP1 (transient overexpression lysate), HeLa whole cell lysate and 3T3 L1 lysate.
TIP47 has been described as cargo protein involved in the trafficking of the mannose-6-phosphate receptor between endosomes and the Golgi complex. Mannose 6-phophate receptors (MPRs) deliver lysosomal hydrolase from the Golgi to endosomes and then return to the Golgi complex. The protein encoded by this gene interacts with the cytoplasmic domains of both cation-independent and cation-dependent MPRs and is required for endosome to Golgi transport. This protein also binds directly to the GTPase RAB9, a member of the RAS oncogene family. The interaction with RAB9 has been shown to increase the affinity of this protein for its cargo. TIP47 has been localized in milk fat globule membranes of human and bovine origin. It has been described as a placental protein, as well. Increased amounts of TIP47 are secreted into circulation of cervix carcinoma patients. Therefore, this protein is probably significant as an oncodevelopmental marker.
Specifications
TIP47 | |
Polyclonal | |
Unconjugated | |
PLIN3 | |
1300012C15Rik; 47 kDa mannose 6-phosphate receptor-binding protein; 47 kDa MPR-binding protein; 47 kDa tail interaction protein; cargo selection protein TIP47; M6prbp1; mannose 6 phosphate receptor binding protein 1; mannose-6-phosphate receptor binding protein 1; mannose-6-phosphate receptor-binding protein 1; MGC11117; MGC2012; perilipin 3; perilipin-3; Placental protein 17; Plin3; PP17; tail-interacting protein, 47 kD; testicular tissue protein Li 114; TIP47; Unknown (protein for MGC:128897) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10226, 595103, 66905 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
TBS with 0.05% sodium azide | |
O60664, Q5BLZ2, Q9DBG5 | |
PLIN3 | |
C-terminus (within residues 350-435) of the human TIP47 protein. [Swiss Prot #O60664]. | |
100 μL | |
Primary | |
Human, Mouse, Monkey, Pig | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.