Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIP47 Polyclonal Antibody, CoraLite488, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc CL48810694100UL
Description
TIP47 Polyclonal antibody specifically detects TIP47 in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
| TIP47 | |
| Polyclonal | |
| CoraLite 488 | |
| O60664 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| M6PRBP1; PLIN3; PP17; TIP47 | |
| Antigen affinity chromatography | |
| RUO | |
| 10226 | |
| -20° C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction