Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAZ2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. 502550794
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-0794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-255-0794 Supplier Invitrogen™ Supplier No. 491037
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BAZ2A Polyclonal Antibody for Western Blot, ChIP

TIP 5 is the large subunit of the nucleolar remodeling complex NoRC. NoRC causes the repression of ribosomal gene transcription. It was demonstrated that histone deacetylation is involved in this repression and that TIP5 is associated with the histone deacetylase HDAC1 and mediates the deacetylation of histones in the vicinity of the rDNA promoter. The interaction of TIP5 and HDAC1, which is necessary for transcriptional repression, is established by the C-terminal PHD finger and bromodomain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAZ2A
Applications ChIP Assay, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with <0.1% sodium azide
Gene BAZ2A
Gene Accession No. Q91YE5, Q9UIF9
Gene Alias AA415431; BAZ2A; bromodomain adjacent to zinc finger domain 2A; bromodomain adjacent to zinc finger domain protein 2A; bromodomain adjacent to zinc finger domain, 2A; C030005G16Rik; C78388; hWALp3; KIAA0314; mKIAA0314; TIP5; Transcription termination factor I-interacting protein 5; TTF-I interacting peptide 5; TTF-I-interacting protein 5; Walp3
Gene Symbols BAZ2A
Host Species Rabbit
Immunogen Recombinant protein TIP5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11176, 116848
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.